Learn More
Thermo Scientific™ Orion™ Powder Chemistry Packs
Product Code. 15159192
Change view
Click to view available options
Type:
Ammonia
Chlorine (free)
Chlorine (total)
Copper (free)
Iron (ferro)
Iron (total)
Manganese
Molybdate/Molybdenum
Nitrite
Phosphate (ortho)
Silica
Sulfate
Range:
0.01 to 0.3 mg/L N
0.01 to 0.8 mg/L N
0.02 to 1.8 mg/L Fe
0.02 to 2 mg/L Cl2
0.02 to 3 mg/L Fe
0.05 to 5 mg/L Cu
0.06 to 2.5 mg/L PO4
0.1 to 18 mg/L Mn
0.5 to 66 mg/L MoO4
1 to 90 mg/L SiO2
5 to 100 mg/L SO4
Unit Size:
100 tests
Each
Pack of 100
Product Code. Type Range unitSize
15159192 Molybdate/Molybdenum 0.5 to 66 mg/L MoO4 Pack of 100
11707727 Ammonia 0.01 to 0.8 mg/L N Each
11717727 Copper (free) 0.05 to 5 mg/L Cu Each
11737727 Nitrite 0.01 to 0.3 mg/L N Each
11747727 Manganese 0.1 to 18 mg/L Mn Each
11712248 Silica 1 to 90 mg/L SiO2 Pack of 100
11757727 Chlorine (free) 0.02 to 2 mg/L Cl2 Pack of 100
11767727 Chlorine (total) 0.02 to 2 mg/L Cl2 Each
11777727 Iron (ferro) 0.02 to 3 mg/L Fe Each
11787727 Iron (total) 0.02 to 1.8 mg/L Fe Each
11797727 Sulfate 5 to 100 mg/L SO4 100 tests
11707737 Phosphate (ortho) 0.06 to 2.5 mg/L PO4 Each
12 options
Product Code. 15159192
Brand: Thermo Scientific™ AC4P42
Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast™ Colorimeters
TRUSTED_SUSTAINABILITY
Specifications
Type Molybdate/Molybdenum
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter
Includes Set of 3: 100 Molybdenum HR1 Powder Packs, 100 Molybdenum HR2 Powder Packs, 100 Molybdenum HR3 Powder Packs
Packaging Powder Pack
Quantity 100 tests
Range 0.5 to 66 mg/L MoO4
Recommended Accessories AQUAfast 24 mm round vials (Cat. No. AC2V24)
Test Method Mercaptoacetic Acid
compliance-icons
Product Identifier
  • Molybdenum HR 1 F10
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : succinic acid, adipic acid
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.