Learn More
Thermo Scientific™ Orion™ Conductivity Standards and Solutions
Product Code. 11707817
Change view
Click to view available options
Packaging:
Bottle
Bottles
Pouch
Conductivity:
100 μS/cm
111.9 mS/cm
12.9 mS
12.9 mS/cm
1413 μS/cm
147 μS/cm
Unit Size:
475mL
Pack of 10
Pack of 5
Product Code. Packaging Conductivity unitSize
11707817 Pouch 1413 μS/cm Pack of 10
10364141 Bottle 100 μS/cm Pack of 5
11906079 Bottle 1413 μS/cm Pack of 5
10075463 Bottles 111.9 mS/cm Pack of 5
11916079 Bottle 12.9 mS/cm Pack of 5
11747717 Bottle 12.9 mS 475mL
11717817 Pouch 147 μS/cm Pack of 10
7 options
Product Code. 11707817
Brand: Thermo Scientific™ 01100710
Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.
Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.
Conductivity Standards
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
TRUSTED_SUSTAINABILITY
Specifications
Description Conductivity Standard
Packaging Pouch
Traceability to NIST Yes
Conductivity 1413 μS/cm
Concentration 692 ppm as NaCl
Color Clear
pH 4.75 to 7.75
Boiling Point 100°C
For Use With (Equipment) Orion pH Meter
Solubility Soluble
Solution Type Standard Solution
Type Conductivity Standard
Quantity 10 x 15 mL
Show More Show Less
Hazard Statement
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.