Learn More
Thermo Scientific™ Orion™ AutoTration™ 500 Autosampler Accessories
Product Code. 11762228
Change view
Click to view available options
Container Size:
150 mL beakers
50 mL beakers
Unit Size:
Pack of 50
Product Code. Container Size unitSize
11762228 50 mL beakers Pack of 50
10150971 150 mL beakers Pack of 50
2 options
Product Code. 11762228
Brand: Thermo Scientific™ ASB050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Maintain automated sample analysis with accessories for our legacy autosampler.
Maintain automated sample analysis with accessories for the legacy Thermo Scientific™ Orion™ AutoTration 500 Autosampler. With a choice of 48- and 28-position racks, this products offers flexibility with interchangeable racks to accommodate sample size and volume.
  • Interchangeable racks to accommodate sample size and volume
  • Choice of 48- and 28-position racks
  • Beaker sizes of 45-, 50- and 150-mL
  • Replacement electrode holders for each model
TRUSTED_SUSTAINABILITY
Specifications
Container Size 50 mL beakers
Description Autosampler 50 mL plastic beakers, 50 pack
Type Titrator Accessories
Includes 50 beakers (50 mL)
For Use With (Equipment) AT5050 AutoTration-500 Autosampler with 48-position rack