Learn More
Thermo Scientific™ Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents
Product Code. 15109202
Change view
Click to view available options
Type:
Ammonia
COD
COD High Range
COD Low Range
COD Mid Range
Fluoride
Manganese
Nitrate
Nitrogen, Total
Phosphate
pH
Range:
0 to 150 mg/L O2
0 to 1500 mg/L O2
0 to 15000 mg/L O2
0.01 to 0.7 mg/L Mn
0.02 to 1.1 mg/L P
0.02 to 1.6 mg/L P
0.02 to 2.5 mg/L N
0.05 to 2 mg/L F
0.06 to 5 mg/L PO4
0.5 to 25 mg/L N
1 to 30 mg/L N
1 to 50 mg/L N
5 to 150 mg/L N
6.5 to 8.4 pH
Unit Size:
25 tests
Each
Pack of 100
Pack of 2
Pack of 25
Pack of 50
Product Code. Type Range unitSize
15109202 Phosphate 0.02 to 1.1 mg/L P Pack of 50
11742218 Fluoride 0.05 to 2 mg/L F Pack of 50
11757757 pH 6.5 to 8.4 pH Pack of 2
15169192 Manganese 0.01 to 0.7 mg/L Mn Pack of 100
15189192 Nitrogen, Total 0.5 to 25 mg/L N Pack of 50
15199192 Nitrogen, Total 5 to 150 mg/L N Pack of 50
15119202 Phosphate 0.02 to 1.6 mg/L P Pack of 50
15179192 Nitrate 1 to 30 mg/L N Pack of 50
15129192 Ammonia 1 to 50 mg/L N Pack of 50
15199182 Ammonia 0.02 to 2.5 mg/L N Pack of 50
15129202 Phosphate 0.06 to 5 mg/L PO4 Pack of 50
11722188 COD 0 to 1500 mg/L O2 25 tests
11742188 COD 0 to 15000 mg/L O2 Pack of 25
11762188 COD 0 to 150 mg/L O2 25 tests
15904012 COD High Range 0 to 15000 mg/L O2 Each
15914012 COD Low Range 0 to 150 mg/L O2 Each
15924012 COD Mid Range 0 to 1500 mg/L O2 Each
17 options
Product Code. 15109202
Brand: Thermo Scientific™ ACD095
Perform colorimetric testing including fluoride, COD, total nitrogen and total phosphate using the Thermo Scientific™ Orion™ AQUAfast Liquid Reagents, Digestion and Reaction Tubes.
Use Thermo Scientific™ Orion™ AQUAfast Digestion Tubes, Reaction Tubes and Liquid Reagents with Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. Digestion and reaction tubes contain the precise amount of reagent, are made of special optical glass and fit the Orion™ AQUAfast COD165 thermoreactor. Digestion tubes measure COD, total nitrogen and total phosphate. Reaction tubes measure ammonia, nitrate and phosphate. Liquid reagents mix quickly and easily in solution and measure fluoride (SPADN), hydrazine and pH.
Changes in nutrient levels can alter the delicate balance in ecosystems and contaminate drinking water. Tube tests make it easy to determine concentrations of species in samples.
Digestion tube tests and reaction tube tests are preprogrammed in the Orion AQ3700 meter. Some of the listed tests have only liquid components, like our AC2009 SPADNS test for fluoride, and some have liquid and powder or liquid and tablet reagents.
TRUSTED_SUSTAINABILITY
Specifications
Description AQUAfast phosphate (total) digestion tubes
EPA Approved Methods Wastewater: Standard Methods 4500-P E-1999
Format Digestion Tube
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ3700 Colorimeter
Includes Set of 4: 50 Phosphate Acid Reagent Digestion Tubes, 50 Potassium Persulfate Powder Packs, 1.54N Sodium Hydroxide Solution, 50 Phosphate Reagent Powder Packs
Quantity 50 Tubes
Range 0.02 to 1.1 mg/L P
Recommended Accessories AQUAfast COD165 Thermoreactor (Cat. No. COD165)
Test Method Persulfate Digestion, Ascorbic Acid
Type Phosphate
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : dipotassium disulphate, potassium antimony(III) oxide tartrate trihydrate