Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
- No. of test: 30
- Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
- Thermo Scientific™ Auto-ID™ eliminates measurement errors
Specifications
Specifications
| Type | Chloride |
| Test Reagent Formats | Ampoule |
| For Use With (Equipment) | Orion AQUAfast AQ4000 Colorimeter |
| Description | AQUAfast chloride Auto-ID ampoules, 30 pack |
| Includes | 30 Auto-ID Ampoules |
| Method | Ferric Thiocyanide |
| For Use With (Application) | Colorimetric water and wastewater testing equipment |
| Recommended Accessories | AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER) |
| Range | 2.5 to 40 mg/L Cl- |
| EPA Approved Methods | Wastewater: Standard Methods 4500-Cl- E-1997 |
| Show More |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.