Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
- No. of test: 30
- Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
- Thermo Scientific™ Auto-ID™ eliminates measurement errors
Spécification
Spécification
| Type | Ammonia |
| Test Reagent Formats | Ampoule |
| For Use With (Equipment) | Orion AQUAfast AQ4000 Colorimeter |
| Description | AQUAfast ammonia HR Auto-ID ampoules, 30 pack |
| Includes | 30 Auto-ID Ampoules |
| Method | Direct Nesslerization |
| For Use With (Application) | Colorimetric water and wastewater testing equipment |
| Recommended Accessories | AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER) |
| Range | 1.5 to 14 mg/L N |
| EPA Approved Methods | Wastewater: ASTM D1426-08 (A) |
| Afficher plus |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.