Learn More
IKA™ Orbital Shaker
GREENER_CHOICE
Product Code. 10316411 Shop All IKA Products
Change view
Click to view available options
Model:
KS 130 Basic
KS 130 Control
KS 260 Basic
KS 260 Control
Load Bearing Capacity (Metric):
2 kg
7.5 kg
Display:
LCD
LED line
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Load Bearing Capacity (Metric) Display unitSize
10316411 KS 130 Basic 2 kg LED line Each
10449792 KS 130 Control 2 kg LED line Each
10706202 KS 260 Basic 7.5 kg LCD Each
10459792 KS 260 Control 7.5 kg LCD Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10316411 Supplier IKA™ Supplier No. 0002980000
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
Accommodates almost all vessel shapes and sizes with a wide range of attachment combinations. IKA™ Orbital Shakers are available in a variety of models with features to serve a number of scientific needs.
KS Orbital shakers come with a wide range of attachment combinations to make it possible to accommodate almost all vessel shapes and sizes. Attachments are not included. Please order separately.
KS 130 models are small, quiet orbital shaker ensures long life with ideal swivel motion, for a maximum shaking weight of 2 kg.
KS 260 models are compact, flat orbital shaker with ideal swivel motion, for a maximum shaking weight of 7.5 kg.
KS 130/260 Basic:
  • Electronic adjustment of speed and timer
  • LED line display shows speed and time adjustment
KS130/260** Control:
  • Electronic adjustment of speed and timer
  • A digital display makes it possible to read the speed, timer function and operating modes
  • Electronic time switching clock: 0 - 9 h 59min or continuous operation (endless)
  • With integrated end point positioning (for automated robot-controlled sampling)
  • All functions can be controlled and documented with labworldsoft™
**Special version with reverse rotating direction on request
KS 501 Digital: Low profile laboratory orbital shaker with a pleasant design, large mounting surface and load capacity of up to 15 kg.
  • Infinitely variable speed control of 0 - 300 rpm
  • Digital display
  • Orbital diameter 30 mm
  • Ideal for vessels with a volume of more than 250mL (round flasks, Erlenmeyer flasks, culture flasks and culture bottles)
  • Ideal for biological and microbiological growth tests, for the production of stock solutions
  • Guaranteed continuous operation (endless) even under extreme loads
  • Includes timer
TRUSTED_SUSTAINABILITY
Specifications
Load Bearing Capacity (Metric) 2 kg
Speed Range 80 to 800 rpm
Temperature Range (Metric) 5°C to 50°C
Wattage 45 W
Product Type Orbital Shaker
Frequency 50/60 Hz
Depth (Metric) 316 mm
Height (Metric) 98 mm
Width (Metric) 270 mm
Weight (Metric) 8.8 kg
Controller Type Analog
Model KS 130 Basic
Display LED line
Timer Range 5 to 50 min.