Learn More
Orbifloxacin, MedChemExpress
Product Code. 16478506
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16478506
Brand: Medchem Express HYB0915
MedChemExpress Orbifloxacin is a synthetic broad-spectrum fluoroquinolone antibiotic which is approved for use in dogs.
Orbifloxacin is a synthetic broad-spectrum fluoroquinolone antibiotic which is approved for use in dogs.
Specifications
Chemical Name or Material Orbifloxacin
Physical Form Solid
CAS 113617-63-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H20F3N3O3
For Use With (Application) COVID-19-immunoregulation
Synonym CP-104354
Solubility Information DMSO : 6 mg/mL (15.18 mM; Need ultrasonic and warming)
SMILES O=C(C1=CN(C2CC2)C3=C(C(F)=C(F)C(N4C[C@H](C)N[C@H](C)C4)=C3F)C1=O)O
Molecular Weight (g/mol) 395.38
Formula Weight 395.38
Percent Purity 99.36%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.