Learn More
Orange/Yellow SantopreneTM Peri-Pump Tubing, 2-Bridge, 0.51 mm (0.02 in.), 6/pack
Click to view available options
Color:
Orange/Yellow
Material:
Santoprene
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Inner | 0.51 mm |
| Material | Santoprene |
| For Use With | Peristaltic Pumps |
| Shape | 2 |
| Color | Orange/Yellow |
| Flexibility | Flexible |
| Quantity | 6/Pk. |