Learn More
Orange/White VitonTM Peri-Pump Tubing, 3-Bridge, 0.64 mm (0.025 in.), 6/pack
Click to view available options
Color:
Orange/White
Material:
Viton
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Inner | 0.64 mm |
| Material | Viton |
| For Use With | Peristaltic Pumps |
| Shape | 3 |
| Color | Orange/White |
| Flexibility | Flexible |
| Quantity | 6/Pk. |