Learn More
OPTIZYME™ 10X Buffer Sau3AI, Fisher BioReagents™
Product Code. 11954862
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11954862 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11954862 Supplier Fisher BioReagents Supplier No. BP80911
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
1X Buffer Composition:
33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate, 0.02% Triton X-100 and 0.1mg/mL BSA
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sau3AI
Physical Form Liquid
Quantity 1 mL
Formulation 33mM Tris Acetate (pH 7.9 at 37°C), 10mM Magnesium Acetate, 66mM Potassium Acetate, 0.1mg/mL BSA, 0.02% Triton™ X-100