Learn More
STEMCELL Technologies OptiPrep™
Product Code. 17109821
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17109821 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17109821 Supplier STEMCELL Technologies Supplier No. 07820
 Restricted Item
Estimated Shipment: 04-11-2026
Density gradient medium for the isolation of viruses, organelles, macromolecules, or cells
  • OptiPrep™ is a density gradient medium recommended for the isolation and purification of DNA or RNA viruses, organelles such as nuclei, mitochondria, endosomes or exosomes, macromolecules, as well as a wide range of cell types
  • OptiPrep™ is a non-ionic iodixanol-based medium with a density of 1.320 ± 0.001 g/mL
  • It has been used clinically in X-ray contrast imaging and has been shown to have lower toxicity compared to ionic gradient compounds such as metrizamide
Specifications
Quantity 250 mL
Cell Type Mononuclear Cells, Other
For Use With (Application) Cell Isolation