Learn More
Hettich™ Optional Lid for Rotors
Click to view available options
For Use With (Equipment):
Hettich Rotors
Unit Size:
Each
Description
- Lid with bio-containment
- For rotors no. 1154-L, 1420-B, 2219-A, 2424-B, 2426-B, 2427-A and 2430-B
- Autoclavable
- Phenol-resistant
Specifications
Specifications
| Type | Biocontainment Lid |
| For Use With (Equipment) | Hettich Rotors |
| Disposable | Reusable |
| Autoclavable | Autoclavable |