Learn More
Macherey-Nagel™ Optima™ 5 MS Accent GC Capillary Column
Product Code. p-8012308 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
12 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.2 mm
0.25 mm
0.32 mm
Film Thickness:
0.2 μm
0.25 μm
0.35 μm
0.5 μm
1 μm
Unit Size:
Each
Product Code. 12422830
missing translation for 'mfr': Macherey-Nagel™ 725815.12
Only null left
Specifications
Diameter (Metric) 0.4 mm
For Use With (Equipment) Ideal for Ion Trap and Qadrupole MS Detectors
Type GC Capillary Column
Diameter (Metric) Inner 0.2 mm
Stationary Phase Optima™ 5 MS Accent
USP Type G1, G2, G38
For Use With (Application) Gas Chromatography (GC)
Temperature Range 320°C to 340°C
Film Thickness 0.35 μm
Length (Metric) 12 m
Quantity 1 Each