Learn More
Macherey-Nagel™ Optima™ 5 HT GC Capillary Column
Product Code. 12758152 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
15 m
30 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
Film Thickness:
0.1 μm
0.25 μm
Unit Size:
Each
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness unitSize
12758152 15 m 0.25 mm 0.25 μm Each
12718152 15 m 0.25 mm 0.1 μm Each
12738152 15 m 0.32 mm 0.1 μm Each
12707710 15 m 0.32 mm 0.25 μm Each
12768152 30 m 0.25 mm 0.25 μm Each
12728152 30 m 0.25 mm 0.1 μm Each
12717710 30 m 0.32 mm 0.25 μm Each
12748152 30 m 0.32 mm 0.1 μm Each
8 options
Product Code. 12758152
Brand: Macherey-Nagel™ 726106.15
Only null left
Specifications
Diameter (Metric) 0.4 mm
For Use With (Equipment) Ideal for MS Detectors, can be Rinsed with Solvents
Type GC Capillary Column
Diameter (Metric) Inner 0.25 mm
Stationary Phase Optima™ 5 HT
USP Type G27, G36
For Use With (Application) Gas Chromatography (GC)
Temperature Range 380°C to 400°C
Film Thickness 0.25 μm
Length (Metric) 15 m
Quantity 1 Each