Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- GC Columns
- GC Capillary Columns
- Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
Product Code. 10776512 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
10 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.2 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.35 μm
0.5 μm
1 μm
Unit Size:
Each
Pack of 50
| Product Code. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | unitSize |
|---|---|---|---|---|
| 10776512 | 30 m | 0.32 mm | 0.35 μm | Each |
| 10506511 | 10 m | 0.25 mm | 0.25 μm | Each |
| 10328071 | 10 m | 0.32 mm | 0.25 μm | Each |
| 10022931 | 10 m | 0.53 mm | 1 μm | Each |
| 10526681 | 15 m | 0.25 mm | 0.25 μm | Each |
| 10671492 | 15 m | 0.32 mm | 0.25 μm | Each |
| 10758622 | 15 m | 0.53 mm | 0.5 μm | Each |
| 10052951 | 15 m | 0.53 mm | 1 μm | Each |
| 10736702 | 25 m | 0.25 mm | 0.25 μm | Each |
| 10419751 | 25 m | 0.2 mm | 0.5 μm | Each |
| 10002911 | 25 m | 0.32 mm | 0.25 μm | Each |
| 10032851 | 25 m | 0.32 mm | 0.35 μm | Each |
| 10526501 | 25 m | 0.32 mm | 0.5 μm | Each |
| 10505551 | 25 m | 0.32 mm | 1 μm | Each |
| 10296751 | 25 m | 0.53 mm | 0.5 μm | Each |
| 10276941 | 30 m | 0.25 mm | 0.25 μm | Each |
| 10012921 | 30 m | 0.25 mm | 0.5 μm | Each |
| 10114711 | 30 m | 0.25 mm | 1 μm | Each |
| 10144561 | 30 m | 0.32 mm | 0.25 μm | Each |
| 10338401 | 25 m | 0.53 mm | 1 μm | Each |
| 10032951 | 30 m | 0.32 mm | 0.5 μm | Each |
| 10429941 | 30 m | 0.32 mm | 1 μm | Each |
| 10681492 | 30 m | 0.53 mm | 0.5 μm | Each |
| 10348401 | 30 m | 0.53 mm | 1 μm | Each |
| 10491272 | 50 m | 0.25 mm | 0.25 μm | Each |
| 10649771 | 50 m | 0.2 mm | 0.5 μm | Each |
| 10420322 | 50 m | 0.32 mm | 0.25 μm | Pack of 50 |
| 10489551 | 50 m | 0.32 mm | 0.35 μm | Each |
| 10768042 | 50 m | 0.32 mm | 0.5 μm | Each |
| 10795932 | 50 m | 0.53 mm | 0.5 μm | Each |
| 10523602 | 50 m | 0.32 mm | 1 μm | Each |
| 10797272 | 60 m | 0.25 mm | 0.5 μm | Each |
| 10516511 | 60 m | 0.25 mm | 0.25 μm | Each |
| 10246941 | 60 m | 0.32 mm | 0.25 μm | Each |
| 10042851 | 60 m | 0.32 mm | 0.35 μm | Each |
| 10246271 | 60 m | 0.32 mm | 0.5 μm | Each |
| 10595381 | 60 m | 0.32 mm | 1 μm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Diameter (Metric) | 0.5 mm |
| Type | High Performance GC Capillary Column |
| Diameter (Metric) Inner | 0.32 mm |
| Stationary Phase | Optima™ 1701 |
| USP Type | G46 |
| For Use With (Application) | Gas Chromatography (GC) |
| Temperature Range | 300°C to 320°C |
| Film Thickness | 0.35 μm |
| Length (Metric) | 30 m |
| Quantity | 1 Each |