Learn More
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
Product Code. 10523602 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
10 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.2 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.35 μm
0.5 μm
1 μm
Unit Size:
Each
Pack of 50
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness unitSize
10523602 50 m 0.32 mm 1 μm Each
10506511 10 m 0.25 mm 0.25 μm Each
10328071 10 m 0.32 mm 0.25 μm Each
10022931 10 m 0.53 mm 1 μm Each
10526681 15 m 0.25 mm 0.25 μm Each
10671492 15 m 0.32 mm 0.25 μm Each
10758622 15 m 0.53 mm 0.5 μm Each
10052951 15 m 0.53 mm 1 μm Each
10736702 25 m 0.25 mm 0.25 μm Each
10419751 25 m 0.2 mm 0.5 μm Each
10002911 25 m 0.32 mm 0.25 μm Each
10032851 25 m 0.32 mm 0.35 μm Each
10526501 25 m 0.32 mm 0.5 μm Each
10505551 25 m 0.32 mm 1 μm Each
10296751 25 m 0.53 mm 0.5 μm Each
10276941 30 m 0.25 mm 0.25 μm Each
10012921 30 m 0.25 mm 0.5 μm Each
10114711 30 m 0.25 mm 1 μm Each
10144561 30 m 0.32 mm 0.25 μm Each
10338401 25 m 0.53 mm 1 μm Each
10776512 30 m 0.32 mm 0.35 μm Each
10032951 30 m 0.32 mm 0.5 μm Each
10429941 30 m 0.32 mm 1 μm Each
10681492 30 m 0.53 mm 0.5 μm Each
10348401 30 m 0.53 mm 1 μm Each
10491272 50 m 0.25 mm 0.25 μm Each
10649771 50 m 0.2 mm 0.5 μm Each
10420322 50 m 0.32 mm 0.25 μm Pack of 50
10489551 50 m 0.32 mm 0.35 μm Each
10768042 50 m 0.32 mm 0.5 μm Each
10795932 50 m 0.53 mm 0.5 μm Each
10797272 60 m 0.25 mm 0.5 μm Each
10516511 60 m 0.25 mm 0.25 μm Each
10246941 60 m 0.32 mm 0.25 μm Each
10042851 60 m 0.32 mm 0.35 μm Each
10246271 60 m 0.32 mm 0.5 μm Each
10595381 60 m 0.32 mm 1 μm Each
37 options
Product Code. 10523602
Brand: Macherey-Nagel™ 726929.50
Only null left
Specifications
Diameter (Metric) 0.5 mm
Type High Performance GC Capillary Column
Diameter (Metric) Inner 0.32 mm
Stationary Phase Optima™ 1701
USP Type G46
For Use With (Application) Gas Chromatography (GC)
Temperature Range 300°C to 320°C
Film Thickness 1 μm
Length (Metric) 50 m
Quantity 1 Each