Learn More
Macherey-Nagel™ Optima™ 1301 GC Capillary Column
Product Code. 10002891 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
1 μm
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness Unit Size
10002891 60 m 0.25 mm 0.25 μm Each
10565551 30 m 0.32 mm 1 μm Each
10194511 50 m 0.25 mm 0.25 μm Each
10378261 25 m 0.32 mm 0.25 μm Each
10412052 25 m 0.25 mm 0.25 μm Each
10144851 50 m 0.32 mm 1 μm Each
10649581 30 m 0.32 mm 0.25 μm Each
10505561 25 m 0.53 mm 1 μm Each
10746142 30 m 0.25 mm 0.25 μm Each
10726522 60 m 0.32 mm 0.25 μm Each
10154851 60 m 0.32 mm 1 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10002891 Supplier Macherey-Nagel™ Supplier No. 726771.60
  • ONLINE EXCLUSIVE PRICE
    12060.00 NOK
    10607.30 NOK / Pack of 1
    Save 1452.70 NOK
    12% Off
Estimated Shipment: 20-11-2026
Specifications
For Use With (Application) Pesticide Analysis
Film Thickness 0.25 μm
Length (Metric) 60 m
Quantity 1 Each
Diameter (Metric) 0.4 mm
USP Type G1, G2, G38
Stationary Phase Optima™ 1301
Type GC Capillary Column
Diameter (Metric) Inner 0.25 mm
Temperature Range (Metric) 300°C to 320°C