Learn More
Macherey-Nagel™ Optima™ 1 MS GC Capillary Column
Product Code. 10155181 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
12 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.2 mm
0.25 mm
0.32 mm
Unit Size:
Each
Product Code. Length (Metric) Diameter (Metric) Inner unitSize
10155181 50 m 0.2 mm Each
10400132 15 m 0.25 mm Each
10124561 30 m 0.25 mm Each
10134561 60 m 0.25 mm Each
10256611 25 m 0.2 mm Each
10409941 30 m 0.32 mm Each
10052871 12 m 0.2 mm Each
10318071 60 m 0.32 mm Each
8 options
Product Code. 10155181
Brand: Macherey-Nagel™ 726201.50
Only null left
Specifications
Diameter (Metric) 0.4 mm
Type GC Capillary Column
Diameter (Metric) Inner 0.2 mm
Stationary Phase Optima™ 1 MS
USP Type G1, G2, G38
For Use With (Application) GC/MS, ECD, General Analyses at Trace Level
Temperature Range 340°C to 360°C
Film Thickness 0.2 μm
Length (Metric) 50 m
Quantity 1 Each