Learn More
Hellma™ Optical Glass Semi-Micro Absorption Cell
Specifications
Specifications
| Type | Semi-micro Absorption Cell |
| Capacity (Metric) | 700 μL |
| Material | Optical Glass |
| Width (Metric) Interior | 4mm |
| Quantity | 2/Pack |
| Path Length | 10 mm |
| Height (Metric) | 45 mm |
| Width (Metric) | 12.5 mm |