Learn More
Hellma™ Optical Glass Micro Cells
Product Code. 10343841
Change view
Click to view available options
Capacity (Metric):
7 mL
17.5 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10343841 7 mL Each
10673711 17.5 mL Each
2 options
Product Code. 10343841
Brand: Hellma™ 60302010
Only null left
Specifications
Type Micro Cell
Capacity (Metric) 7 mL
Material Optical Glass
Closure Material PTFE
Width (Metric) Interior 9.5mm
Quantity 1Each
Path Length 20 mm
Wavelength Range 360 to 2500 nm
Height (Metric) 45 mm
Width (Metric) 12.5 mm