Learn More
Hellma™ Optical Glass High Precision Macro Absorption Cells With PTFE Stopper
Product Code. 10737381
Click to view available options
Capacity (Metric):
1750 μL
3500 μL
7000 μL
Unit Size:
Each
Pack of 2
Product Code. 10737381
Brand: Hellma™ 1101020
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type Macro Absorption Cell
Capacity (Metric) 3500 μL
Material Optical Glass
Closure Material PTFE
Width (Metric) Interior 9.5mm
Path Length 10 mm
Wavelength Range 320 to 2500 nm
Height (Metric) 46 mm
Width (Metric) 12.5 mm
For Use With (Application) Photometric and Colorimetric Measurements