Learn More
Marienfeld Superior™ Cover Glasses for Counting Chambers and Haemacytometers
Description
- Made of special optical glass
- Ready for use
- Ground and polished edges
- Additional feature: The excellent quality of the edges make the haemacytometer cover glasses well suitable for blood smears
- Individually packed in poly bags, 10 poly bags in a plastic box, 10 plastic boxes in a cardboard box
- For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark and batch number for comprehensive information and traceability
Specifications
Specifications
| Product Type | Coverslip |
| Thickness (Metric) | 0.4 mm |
| Dimensions (L x W) | 20 x 26 mm |
| Description | Ground and Polished Edges |
| For Use With (Equipment) | Counting Chambers and Haemacytometers |
| Quantity | 100 |