Learn More
Gibco™ Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement
GREENER_CHOICE
Product Code. 11574506
Change view
Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
500mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11574506 10 x 500 mL Pack of 10
11564506 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11574506 Supplier Gibco™ Supplier No. 51985042
Simplify Your Gene Expression Workflow
Use promo code 27975
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement
Opti-MEM™ Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change to growth rate or morphology. Opti-MEM™ is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Gibco™ Opti-MEM™ modifications for a range of cell culture applications. The complete formulation is confidential. For more information, please contact Technical Services.
Gibco™ Opti-MEM™ is a unique media that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Gibco™ Opti-MEM™ with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ Opti-MEM™ is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Opti-MEM™ product made in our Grand Island facility (31985-070). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Gibco™ Opti-MEM™ uses a sodium bicarbonate buffer system (2.4 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1 X
Form Liquid
Product Type Reduced-Serum Medium
With Additives GlutaMAX, Phenol Red, Sodium Pyruvate
For Use With (Application) Mammalian cell culture, transfection
Quantity 10 x 500 mL
Certifications/Compliance ISO 13485 standards, FDA
pH 7.0 to 7.4
Content And Storage 2 to 8°C
Description Opti-Mem™ Reduced-Serum Medium, 10 x 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With Glutamax™, Phenol Red, Sodium Pyruvate, 18 Months Shelf Life, 280 to 320mOsm/kg Osmolality, For Mammalian Cell Culture, Transfection
For In Vitro Diagnostic Use.