Learn More
Hirschmann™ openend™ Disposable Capillary Pipettes, Na-hep
Product Code. 10329901
Change view
Click to view available options
Volume (Metric):
20 μL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10329901 20 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10329901 Supplier Hirschmann™ Supplier No. 905022001
  • ONLINE EXCLUSIVE PRICE
    2245.00 NOK
    1649.53 NOK / Pack of 1000
    Save 595.47 NOK
    26% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ openend™ Disposable Capillary Pipettes can be utilized for a wide range of scientific and medical applications.
  • Sodium Heparinized
  • Conformity certified
  • Manufactured from durable DURAN™ glass
  • CE acc. IVD 98/79 EC
Specifications
Accuracy 0.5%
Certifications/Compliance CE, IVD 98/79 EC
Material Glass
Volume (Metric) 20 μL
Length (Metric) 60 ±0.5 mm
Type Capillary Pipette