Learn More
Thermo Scientific™ Open-Top Screw Caps
Product Code. 11597350 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
3 mL
3.5 mL
7 mL
10 mL
14 mL
Unit Size:
Pack of 72
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11597350 10 mL Pack of 72
11571024 3.5 mL Pack of 72
11567350 7 mL Pack of 72
11577350 14 mL Pack of 72
11501034 3 mL Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11597350 Supplier Thermo Scientific™ Supplier No. TS13219
Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.
Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.
  • Fit into Reacti-Vial™ small reaction vials
  • Used with PTFE/Rubber laminated discs
TRUSTED_SUSTAINABILITY
Specifications
Description TS-13219
Volume (Metric) 10 mL
Closure Size 24 mm
For Use With (Equipment) 10 mL Reacti-Vials and 40 mL Screw Cap Septum Vials
Product Type Open Top Screw Cap