Learn More
Thermo Scientific™ Open-Top Screw Caps
Product Code. 11577350 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
3 mL
3.5 mL
7 mL
10 mL
14 mL
Unit Size:
Pack of 72
Product Code. Volume (Metric) unitSize
11577350 14 mL Pack of 72
11571024 3.5 mL Pack of 72
11567350 7 mL Pack of 72
11501034 3 mL Pack of 72
11597350 10 mL Pack of 72
5 options
Product Code. 11577350
Brand: Thermo Scientific™ TS13217
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.
Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.
  • Fit into Reacti-Vial™ small reaction vials
  • Used with PTFE/Rubber laminated discs
TRUSTED_SUSTAINABILITY
Specifications
Description TS-13217
Volume (Metric) 14 mL
Closure Size 18 mm
For Use With (Equipment) 14 mL Screw Cap Septum Vials
Product Type Open Top Screw Cap