Learn More
Köttermann Open Tall Storage Cabinet
Product Code. 17392079
Change view
Click to view available options
No. of Drawers:
4
None
No. of Shelves:
4
None
Dimensions (D x W x H):
350 x 600 x 1920 mm
350 x 900 x 1920 mm
500 x 600 x 1920 mm
500 x 900 x 1920 mm
350 x 1200 x 1920 mm
500 x 1200 x 1920 mm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Drawers No. of Shelves Dimensions (D x W x H) Unit Size
17392079 None 4 350 x 600 x 1920 mm Each
17372089 None 4 350 x 900 x 1920 mm Each
17352079 None 4 350 x 1200 x 1920 mm Each
17342089 None 4 500 x 600 x 1920 mm Each
17332089 4 None 500 x 600 x 1920 mm Each
17302099 None 4 500 x 900 x 1920 mm Each
17382079 None 4 500 x 1200 x 1920 mm Each
17372079 4 None 500 x 1200 x 1920 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17392079 Supplier Köttermann Supplier No. 30100033
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
The open-front design of this Open Tall Storage Cabinet provides easy access to stored equipment and utensils.
  • Tall, slender design expands storage with a small footprint
  • Open front provides easy access to stored items
Specifications
Color White
Adjustable No
Cabinet Type Tall Storage
Dimensions (D x W x H) 350 x 600 x 1920 mm
Door Type None
Finish Epoxy Coated
For Use With (Application) Storage
No. of Doors None
No. of Drawers None
No. of Shelves 4
Warranty None
Weight (Metric) 46.4 kg
With Casters No
Material Steel