Learn More
Köttermann Open Tall Storage Cabinet
The open-front design of this Open Tall Storage Cabinet provides easy access to stored equipment and utensils.
Brand: Köttermann 301-00069
Additional Details : Weight : 59.83950kg
Description
- Tall, slender design expands storage with a small footprint
- Open front provides easy access to stored items
Specifications
None | |
Storage | |
No | |
59.9 kg | |
4 | |
None | |
Steel |
None | |
No | |
350 x 900 x 1920 mm | |
None | |
Epoxy Coated | |
White |