Learn More
Köttermann Open Mobile Cabinet
Product Code. 17322129
Click to view available options
No. of Drawers:
2
3
None
No. of Shelves:
1
None
Dimensions (D x W x H):
500 x 450 x 590 mm
500 x 450 x 740 mm
500 x 600 x 590 mm
500 x 600 x 740 mm
500 x 900 x 590 mm
500 x 900 x 740 mm
516 x 450 x 590 mm
516 x 600 x 590 mm
500 x 1200 x 590 mm
500 x 1200 x 740 mm
Unit Size:
Each
Product Code. 17322129
Brand: Köttermann 31500133
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Open Mobile Cabinet offers versatile storage space with an open-front design.
  • Versatile storage space
  • Mobile with lockable wheels
  • Open-front design provides easy access to stored items
  • Many sizes and configurations available
Specifications
Color White
Cabinet Type Mobile
Dimensions (D x W x H) 500 x 900 x 740 mm
Door Type None
Finish Epoxy Coated
Frame Material Steel
No. of Doors None
No. of Drawers None
No. of Shelves None
Warranty None
Wheel Type Plastic with Brakes
With Casters Yes
Material Steel