Learn More
Masterflex™ Opaque Black Viton 2-stopped Pump Tubing
Brand: Masterflex™ 97628-30
Additional Details : Weight : 0.80000kg
Specifications
2-stop Tubing | |
Viton | |
-13°C | |
Acids, Organic Solvents | |
Opaque Black |
1.14 mm | |
Acid and solvent transfer, high-temperature work | |
12 Pack | |
0.017 to 38mL/min. | |
-13°C to +204°C |