Learn More
Kolmi Hopen™ Op-Air™ PRO Oxygen Type IIR FFP1 Respiratory Mask
This respiratory pouch mask is ideal for protection against infectious diseases and in dusty environments.
Brand: Kolmi Hopen™ M51010S-WH
Additional Details : Weight : 4.56000kg
Description
- Filtering half mask with FFP1 NR D efficiency class
- Developed to protect against solid and liquid aerosols and particles
- Protects environment against droplets emitted by wearer and protects wearer from potential body fluid splashes
- Knitted head loops are ultrasonically welded for optimal comfort
- Fits perfectly to face to prevent exposure
- Made in France
Specifications
(EU) 2017/745, 2016/425, EN 14683:2019, EN 149:2001+A1:2009, ISO 9001, ISO 13485 | |
Pouch/Duckbill | |
Polypropylene | |
Yes |
White | |
Single-use | |
Medium | |
50 Pk. |