Learn More
Kolmi™ Op Air Pro Oxygen FFP3 NR D Mass Balance Masks
GREENER_CHOICE
Product Code. 17724799 Shop All Medicom Products
Click to view available options
Size:
Medium
Large
Unit Size:
Pack of 50
Product Code. 17724799
Brand: Kolmi™ M53114SWHMB
Only null left
Add to basket
Add to basket
Op Air Pro Oxygen FFP3 masks are easy to put on with high breathability due to high quality raw material. Made with the Mass Balance approach from recycled materials.
  • Filtering half mask, efficiency class FFP3 NR D developed to protect against solid and liquid aerosols and particles
  • Type IIR certified, it protects the environment against droplets emitted by the wearer and it protects from potential body fluid splashes
  • Knitted headloops ultrasonically welded for optimal comfort
  • Fits perfectly to face to limit inward leakage thanks to its shape and soft materials
  • Hypoallergenic and has been dermatologically tested
  • Packaged in individual polybags
  • Made in France
Specifications
Certifications/Compliance EN 149: 2001 + A1: 2009, DM Class I, EN 14683: 2019 + AC: 2019
Color White
Size Large
For Use With (Application) Industry
Style Pouch