Learn More
Gibco™ OneFeed™ Supplement
A versatile medium supplement.
Brand: Gibco™ 670109
Additional Details : Weight : 0.40400kg
Description
Gibco™ OneFeed™ Supplement is a chemically-defined, animal-free, protein-free feed shown to enhance protein production when used in a variety of commercially available medium.Specifications
Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers. | |
Animal Origin-free, Chemically-defined, Protein-free | |
Media Supplement | |
10 L |
CHO DG44, BHK-21, CHO-K1, CHO | |
Powder | |
Container | |
Animal origin-free |