Learn More
Hamilton™ One-Hub Assembled Tubing Assemblies
Click to view available options
Needle Gauge:
8 G
10 G
11 G
12 G
13 G
14 G
16 G
17 G
19 G
20 G
26 G
30 G
Length (Metric):
304.8 mm
Custom
Unit Size:
Each
Description
- Fitting manufactured from CTFE
- Tubing manufactured from PTFE
- For female luers
- A variety of gauges and lengths available
Specifications
Specifications
| Type | Tubing Assembly |
| Connections | Female Luer |
| Quantity | 1/Pk. |
| Material | CTFE (Fitting), PTFE (Tubing) |
| For Use With (Equipment) | 26 ga. Connections |
| Length (Metric) | Custom |
| Needle Gauge | 26 G |