Learn More
Hamilton™ One-Hub Assembled Tubing Assemblies
Product Code. 11503084
Change view
Click to view available options
Needle Gauge:
8 G
10 G
11 G
12 G
13 G
14 G
16 G
17 G
19 G
20 G
26 G
30 G
Length (Metric):
304.8 mm
Custom
Unit Size:
Each
Product Code. Needle Gauge Length (Metric) unitSize
11503084 8 G 304.8 mm Each
10394703 11 G 304.8 mm Each
10106833 10 G 304.8 mm Each
11543084 12 G 304.8 mm Each
10170883 13 G 304.8 mm Each
11573124 20 G Custom Each
11583124 20 G Custom Each
11563084 14 G 304.8 mm Each
11563124 19 G Custom Each
11593134 12 G Custom Each
11593084 17 G 304.8 mm Each
11503134 26 G Custom Each
11523134 30 G Custom Each
11533144 16 G Custom Each
14 options
Product Code. 11503084
Brand: Hamilton™ 90608
Providing a variety of connection solutions in the laboratory.
  • Fitting manufactured from CTFE
  • Tubing manufactured from PTFE
  • For female luers
  • A variety of gauges and lengths available
Specifications
Type Tubing Assembly
Connections Female Luer
Quantity 1/Pk.
Material CTFE (Fitting), PTFE (Tubing)
For Use With (Equipment) 8 ga. Connections
Length (Metric) 304.8 mm
Needle Gauge 8 G