Learn More
Hamilton™ One-Hub Assembled Tubing Assemblies
Product Code. 10394703
Click to view available options
Needle Gauge:
8 G
10 G
11 G
12 G
13 G
14 G
16 G
17 G
19 G
20 G
26 G
30 G
Length (Metric):
304.8 mm
Custom
Unit Size:
Each
Product Code. 10394703
Brand: Hamilton™ 90611
Providing a variety of connection solutions in the laboratory.
  • Fitting manufactured from CTFE
  • Tubing manufactured from PTFE
  • For female luers
  • A variety of gauges and lengths available
Specifications
Type Tubing
Material PTFE
Length (Metric) 304.8 mm
Needle Gauge 11 G