Learn More
Buerkle™ LiquiSampler
Product Code. 10759222 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
150 mL
250 mL
500 mL
Material:
PTFE, FEP
Polypropylene
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Material Unit Size
10759222 150 mL Polypropylene Each
10177581 150 mL PTFE, FEP Each
10115360 250 mL Polypropylene Each
10300061 250 mL PTFE, FEP Each
10637101 500 mL Polypropylene Each
10155310 500 mL PTFE, FEP Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10759222 Supplier Buerkle™ Supplier No. 53301060
Estimated Shipment: 23-12-2026
Buerkle™ LiquiSampler for liquids, according to DIN EN ISO 15528. Made of ultra pure and chemically inert PTFE/FEP or PP for contamination-free sampling. Sampling from open and closed containers, barrels, tanks, silos, water courses. Suitable for all openings of at least 32 mm in diameter.
  • Practical one-handed use at the push of a button
  • With scraping ring for marking of depth and cleaning
  • Transparent
  • Available in PTFE/FEP or PP transparent
  • Point sampling, all-layer sampling and bottom sampling possible
  • Suitable for all commercially available barrels and containers with opening of at least 32 mm in diameter
  • Easy to clean
TRUSTED_SUSTAINABILITY
Specifications
Product Type LiquiSampler
Material Polypropylene
Autoclavable Not Autoclavable
Includes Scraping Ring
Handle Type Button - one-handed use
Capacity (English) 5.07 oz.
Capacity (Metric) 150 mL
Color Transparent
Certifications/Compliance DIN EN ISO 15528
For Use With (Application) Sampling From Open and Closed Containers, Barrels, Tanks, Silos, Water Courses, Point Sampling, All-Layer and Soil Sampling, Suitable for Use in Foodstuffs, for Free-flowing material
Diameter (English) Opening 0.67 in.
Diameter (Metric) Opening 17 mm
Diameter (Metric) Outer 32 mm
Length (English) 29.06 in.
Length (Metric) 738 mm
Length (Metric) Shaft 60 cm