Learn More
Köttermann One Door Tall Storage Cabinet
Product Code. 17312089
Change view
Click to view available options
Hinge Side:
Left
Right
No. of Shelves:
3
4
Dimensions (D x W x H):
366 x 600 x 1920 mm
516 x 600 x 1920 mm
Unit Size:
Each
Product Code. Hinge Side No. of Shelves Dimensions (D x W x H) unitSize
17312089 Right 3 366 x 600 x 1920 mm Each
17302089 Left 4 366 x 600 x 1920 mm Each
17322089 Right 4 366 x 600 x 1920 mm Each
17352089 Right 4 516 x 600 x 1920 mm Each
4 options
Product Code. 17312089
Brand: Köttermann 30100043
This item requires a price update prior to being ordered.
Request a Price for this item here.
This One Door Tall Storage Cabinet features a single door to protect stored equipment and utensils.
  • Tall, slender design expands storage with a small footprint
  • Single door protects stored items
Specifications
Color White
Adjustable No
Cabinet Type Tall Storage
Dimensions (D x W x H) 366 x 600 x 1920 mm
Door Type Hinged
Finish Epoxy Coated
For Use With (Application) Storage
Hinge Side Right
Locking Mechanism Yes
No. of Doors 1
No. of Drawers None
No. of Shelves 3
Warranty None
Weight (Metric) 60.3 kg
With Casters No
Material Steel