Learn More
Ondansetron, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Ondansetron |
| CAS | 99614-02-5 |
| Analyte or Component Names | Ondansetron |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H19 N3 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ondansetron, 1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one, 1,2,3,4-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-9H-carbazol-4-one, GR 38032, GR 38032X, Ondanles, Setron, Yatrox, Zofran, Zophren, Zudan |
| InChI Formula | InChI=1S/C18H19N3O/c1-12-19-9-10-21(12)11-13-7-8-16-17(18(13)22)14-5-3-4-6-15(14)20(16)2/h3-6,9-10,13H,7-8,11H2,1-2H3 |
| Solution Type | Neat |
| SMILES | Cc1nccn1CC2CCc3c(C2=O)c4ccccc4n3C |
| Show More |