Learn More
Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns
Product Code. 15895294
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Length (Metric):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Unit Size:
Each
Product Code. Diameter (Metric) Inner Length (Metric) unitSize
15895294 25 mm 330 mm Each
15885274 6.6 mm 50 mm Each
15895274 6.6 mm 100 mm Each
15805284 6.6 mm 150 mm Each
15815284 6.6 mm 250 mm Each
15825284 6.6 mm 330 mm Each
15835284 6.6 mm 400 mm Each
15845284 10 mm 100 mm Each
15855284 10 mm 150 mm Each
15865284 10 mm 250 mm Each
15875284 10 mm 330 mm Each
15885284 10 mm 400 mm Each
15895284 10 mm 500 mm Each
15805294 15 mm 100 mm Each
15815294 15 mm 150 mm Each
15825294 15 mm 250 mm Each
15835294 15 mm 330 mm Each
15845294 15 mm 400 mm Each
15855294 15 mm 500 mm Each
15865294 25 mm 100 mm Each
15875294 25 mm 150 mm Each
15885294 25 mm 250 mm Each
15805304 25 mm 400 mm Each
15815304 25 mm 500 mm Each
15825304 35 mm 150 mm Each
15835304 35 mm 250 mm Each
15845304 35 mm 330 mm Each
15855304 35 mm 400 mm Each
15865304 50 mm 250 mm Each
15875304 50 mm 330 mm Each
15885304 50 mm 400 mm Each
15895304 50 mm 500 mm Each
32 options
Product Code. 15895294
Brand: Diba 006ERG2533
This item requires a price update prior to being ordered.
Request a Price for this item here.
Mix and match your Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Column lengths while using your existing endpieces for ultimate flexibility
Use a different length tube with your existing endpieces to meet your specific requirements.
Specifications
For Use With (Equipment) Omnifit™ EZ and EZ SolventPlus™ Columns
Diameter (Metric) Inner 25 mm
Type Liquid Chromatography Column
Length (Metric) 330 mm
Material Glass