Learn More
Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns
Click to view available options
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Length (Metric):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Unit Size:
Each
Description
Use a different length tube with your existing endpieces to meet your specific requirements.
Specifications
Specifications
| For Use With (Equipment) | Omnifit™ EZ and EZ SolventPlus™ Columns |
| Diameter (Metric) Inner | 35 mm |
| Type | Liquid Chromatography Column |
| Length (Metric) | 400 mm |
| Material | Glass |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.