Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- Low to Medium Pressure LC Columns
- Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns
Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns
Product Code. 15815294
| Product Code. | Diameter (Metric) Inner | Length (Metric) | unitSize |
|---|---|---|---|
| 15815294 | 15 mm | 150 mm | Each |
| 15885274 | 6.6 mm | 50 mm | Each |
| 15895274 | 6.6 mm | 100 mm | Each |
| 15805284 | 6.6 mm | 150 mm | Each |
| 15815284 | 6.6 mm | 250 mm | Each |
| 15825284 | 6.6 mm | 330 mm | Each |
| 15835284 | 6.6 mm | 400 mm | Each |
| 15845284 | 10 mm | 100 mm | Each |
| 15855284 | 10 mm | 150 mm | Each |
| 15865284 | 10 mm | 250 mm | Each |
| 15875284 | 10 mm | 330 mm | Each |
| 15885284 | 10 mm | 400 mm | Each |
| 15895284 | 10 mm | 500 mm | Each |
| 15805294 | 15 mm | 100 mm | Each |
| 15825294 | 15 mm | 250 mm | Each |
| 15835294 | 15 mm | 330 mm | Each |
| 15845294 | 15 mm | 400 mm | Each |
| 15855294 | 15 mm | 500 mm | Each |
| 15865294 | 25 mm | 100 mm | Each |
| 15875294 | 25 mm | 150 mm | Each |
| 15885294 | 25 mm | 250 mm | Each |
| 15895294 | 25 mm | 330 mm | Each |
| 15805304 | 25 mm | 400 mm | Each |
| 15815304 | 25 mm | 500 mm | Each |
| 15825304 | 35 mm | 150 mm | Each |
| 15835304 | 35 mm | 250 mm | Each |
| 15845304 | 35 mm | 330 mm | Each |
| 15855304 | 35 mm | 400 mm | Each |
| 15865304 | 50 mm | 250 mm | Each |
| 15875304 | 50 mm | 330 mm | Each |
| 15885304 | 50 mm | 400 mm | Each |
| 15895304 | 50 mm | 500 mm | Each |
Product Code. 15815294
Brand: Diba 006ERG1515
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Specifications
Specifications
| For Use With (Equipment) | Omnifit™ EZ and EZ SolventPlus™ Columns |
| Diameter (Metric) Inner | 15 mm |
| Type | Liquid Chromatography Column |
| Length (Metric) | 150 mm |
| Material | Glass |