Learn More
Diba Omnifit™ Replacement Endpieces for EZ Columns
Product Code. 15815274
Encompass_Preferred
Change view
Click to view available options
Endcapped:
Adjustable
Fixed
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Unit Size:
Each
Product Code. Endcapped Diameter (Metric) Inner unitSize
15815274 Fixed 50 mm Each
15805264 Adjustable 6.6 mm Each
15815264 Adjustable 10 mm Each
15825264 Adjustable 15 mm Each
15835264 Adjustable 25 mm Each
15845264 Adjustable 35 mm Each
15855264 Adjustable 50 mm Each
15865264 Fixed 6.6 mm Each
15875264 Fixed 10 mm Each
15885264 Fixed 15 mm Each
15895264 Fixed 25 mm Each
15805274 Fixed 35 mm Each
12 options
Product Code. 15815274
Brand: Diba 006EFE50
This item requires a price update prior to being ordered.
Request a Price for this item here.
Select from fixed or adjustable Omnifit™ Replacement Endpieces for EZ Columns
Specifications
Endcapped Fixed
For Use With (Equipment) Omnifit™ EZ Columns
Diameter (Metric) Inner 50 mm
Type Liquid Chromatography Column
Length (Metric) 4 cm
Material PTFE