Learn More
Diba Omnifit™ O-Rings for Chromatography Columns
Product Code. 15835564
Encompass_Preferred
Click to view available options
Diameter (Metric) Inner:
3 mm
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Material:
FFKM
FKM
Unit Size:
Pack of 10
Pack of 2
Pack of 6
Product Code. 15835564
Brand: Diba 006OR03V
This item requires a price update prior to being ordered.
Request a Price for this item here.
Select from FKM or FFKM (Chemraz™) Omnifit™ O-Rings for Chromatography Columns
Specifications
For Use With (Equipment) Omnifit™ HiT Chromatography Columns
Quantity 6 Pk.
Diameter (Metric) Inner 3 mm
Type Liquid Chromatography Column
Length (Metric) 3 mm
Material FKM
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.