Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- Low to Medium Pressure LC Columns
- Diba Omnifit™ Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns
Diba Omnifit™ Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns
Product Code. 15885514
| Product Code. | Diameter (Metric) Inner | Material | Pore Size | unitSize |
|---|---|---|---|---|
| 15885514 | 6.6 mm | PTFE | 30 μm | Pack of 20 |
| 15845514 | 6.6 mm | PTFE | 5 μm | Pack of 20 |
| 15855514 | 6.6 mm | PTFE | 10 μm | Pack of 20 |
| 15865514 | 6.6 mm | PTFE | 20 μm | Pack of 20 |
| 15875514 | 6.6 mm | Polyethylene | 25 μm | Pack of 20 |
| 15895514 | 6.6 mm | PTFE | 50 μm | Pack of 20 |
| 15805524 | 10 mm | PTFE | 5 μm | Pack of 20 |
| 15815524 | 10 mm | PTFE | 10 μm | Pack of 20 |
| 15825524 | 10 mm | PTFE | 20 μm | Pack of 20 |
| 15835524 | 10 mm | Polyethylene | 25 μm | Pack of 20 |
| 15845524 | 10 mm | PTFE | 30 μm | Pack of 20 |
| 15855524 | 10 mm | PTFE | 50 μm | Pack of 20 |
| 15885524 | 15 mm | PTFE | 20 μm | Pack of 20 |
| 15805534 | 15 mm | PTFE | 30 μm | Pack of 20 |
| 15815534 | 15 mm | PTFE | 50 μm | Pack of 20 |
| 15825534 | 25 mm | PTFE | 5 μm | Pack of 20 |
| 15835534 | 25 mm | PTFE | 10 μm | Pack of 20 |
| 15845534 | 25 mm | PTFE | 20 μm | Pack of 20 |
| 15855534 | 25 mm | Polyethylene | 25 μm | Pack of 20 |
| 15865534 | 25 mm | PTFE | 30 μm | Pack of 20 |
| 15875534 | 25 mm | PTFE | 50 μm | Pack of 20 |
| 15805544 | 35 mm | PTFE | 20 μm | Pack of 20 |
| 15825544 | 35 mm | PTFE | 30 μm | Pack of 20 |
| 15835544 | 35 mm | PTFE | 50 μm | Pack of 20 |
| 15865544 | 50 mm | PTFE | 20 μm | Pack of 20 |
| 15885544 | 50 mm | PTFE | 30 μm | Pack of 20 |
| 15895544 | 50 mm | PTFE | 50 μm | Pack of 20 |
Product Code. 15885514
Brand: Diba 006FR0630
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Specifications
| For Use With (Equipment) | Omnifit™ EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns |
| Diameter (Metric) Inner | 6.6 mm |
| Type | Liquid Chromatography Column |
| Pore Size | 30 μm |
| Length (Metric) | 6 mm |
| Material | PTFE |