Learn More
Diba Omnifit™ EZ Chromatography Columns with One Fixed and One Adjustable Endpieces
Product Code. 15895324
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Column Capacity:
0.1 to 0.7 mL
0.1 to 2.4 mL
0.2 to 5.5 mL
0.4 to 12.4 mL
1.2 to 34.4 mL
1.4 to 4.1 mL
108.0 to 147.3 mL
11.0 to 17.3 mL
134.7 to 211.7 mL
142.4 to 181.6 mL
17.3 to 23.6 mL
19.6 to 58.9 mL
191.4 to 230.7 mL
211.7 to 288.6 mL
22.8 to 29.1 mL
24.7 to 38.9 mL
279.0 to 356.0 mL
280.4 to 440.7 mL
3.1 to 9.4 mL
30.6 to 36.9 mL
38.5 to 115.5 mL
38.9 to 53.0 mL
4.8 to 7.5 mL
440.7 to 600.9 mL
51.2 to 65.4 mL
580.9 to 741.1 mL
68.7 to 108.0 mL
68.9 to 83.1 mL
7.1 to 21.2 mL
7.5 to 10.3 mL
781.2 to 941.4 mL
9.9 to 12.7 mL
Unit Size:
Each
Product Code. Length (Metric) Diameter (Metric) Inner Column Capacity unitSize
15895324 150 mm 6.6 mm 1.4 to 4.1 mL Each
15835324 50 mm 6.6 mm 0.1 to 0.7 mL Each
15865324 100 mm 6.6 mm 0.1 to 2.4 mL Each
15825334 250 mm 6.6 mm 4.8 to 7.5 mL Each
15855334 330 mm 6.6 mm 7.5 to 10.3 mL Each
15885334 400 mm 6.6 mm 9.9 to 12.7 mL Each
15815344 100 mm 10 mm 0.2 to 5.5 mL Each
15845344 150 mm 10 mm 3.1 to 9.4 mL Each
15875344 250 mm 10 mm 11.0 to 17.3 mL Each
15805354 330 mm 10 mm 17.3 to 23.6 mL Each
15835354 400 mm 10 mm 22.8 to 29.1 mL Each
15865354 500 mm 10 mm 30.6 to 36.9 mL Each
15895354 100 mm 15 mm 0.4 to 12.4 mL Each
15825364 150 mm 15 mm 7.1 to 21.2 mL Each
15855364 250 mm 15 mm 24.7 to 38.9 mL Each
15885364 330 mm 15 mm 38.9 to 53.0 mL Each
15815374 400 mm 15 mm 51.2 to 65.4 mL Each
15845374 500 mm 15 mm 68.9 to 83.1 mL Each
15875374 100 mm 25 mm 1.2 to 34.4 mL Each
15805384 150 mm 25 mm 19.6 to 58.9 mL Each
15835384 250 mm 25 mm 68.7 to 108.0 mL Each
15865384 330 mm 25 mm 108.0 to 147.3 mL Each
15895384 400 mm 25 mm 142.4 to 181.6 mL Each
15825394 500 mm 25 mm 191.4 to 230.7 mL Each
15855394 150 mm 35 mm 38.5 to 115.5 mL Each
15885394 250 mm 35 mm 134.7 to 211.7 mL Each
15815404 330 mm 35 mm 211.7 to 288.6 mL Each
15845404 400 mm 35 mm 279.0 to 356.0 mL Each
15875404 250 mm 50 mm 280.4 to 440.7 mL Each
15805414 330 mm 50 mm 440.7 to 600.9 mL Each
15835414 400 mm 50 mm 580.9 to 741.1 mL Each
15865414 500 mm 50 mm 781.2 to 941.4 mL Each
32 options
Product Code. 15895324
Brand: Diba 006EZ0615AF
This item requires a price update prior to being ordered.
Request a Price for this item here.
Includes
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting.
Omnifit™ EZ Chromatography Columns with One Fixed and One Adjustable Endpieces are ergonomically designed endpieces provide simple and quick height adjustment
  • PEEK™ column threads offer outstanding durability
  • Precision borosilicate glass columns provide inertness, visibility, and performance
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Specifications
Temperature (Metric) Range 4 to 40°C
Diameter (Metric) Inner 6.6 mm
Type Liquid Chromatography Column
Max. Pressure 900 psi
Pore Size 10 and 30 μm
Column Capacity 1.4 to 4.1 mL
Length (Metric) 150 mm
Material Borosilicate Glass, PTFE, Polyethylene, FKM/FPM
Max. Temperature 40°C
Includes Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting.