Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- Low to Medium Pressure LC Columns
- Diba Omnifit™ EZ Chromatography Columns with One Fixed and One Adjustable Endpieces
Diba Omnifit™ EZ Chromatography Columns with One Fixed and One Adjustable Endpieces
Product Code. 15825334
| Product Code. | Length (Metric) | Diameter (Metric) Inner | Column Capacity | unitSize |
|---|---|---|---|---|
| 15825334 | 250 mm | 6.6 mm | 4.8 to 7.5 mL | Each |
| 15835324 | 50 mm | 6.6 mm | 0.1 to 0.7 mL | Each |
| 15865324 | 100 mm | 6.6 mm | 0.1 to 2.4 mL | Each |
| 15895324 | 150 mm | 6.6 mm | 1.4 to 4.1 mL | Each |
| 15855334 | 330 mm | 6.6 mm | 7.5 to 10.3 mL | Each |
| 15885334 | 400 mm | 6.6 mm | 9.9 to 12.7 mL | Each |
| 15815344 | 100 mm | 10 mm | 0.2 to 5.5 mL | Each |
| 15845344 | 150 mm | 10 mm | 3.1 to 9.4 mL | Each |
| 15875344 | 250 mm | 10 mm | 11.0 to 17.3 mL | Each |
| 15805354 | 330 mm | 10 mm | 17.3 to 23.6 mL | Each |
| 15835354 | 400 mm | 10 mm | 22.8 to 29.1 mL | Each |
| 15865354 | 500 mm | 10 mm | 30.6 to 36.9 mL | Each |
| 15895354 | 100 mm | 15 mm | 0.4 to 12.4 mL | Each |
| 15825364 | 150 mm | 15 mm | 7.1 to 21.2 mL | Each |
| 15855364 | 250 mm | 15 mm | 24.7 to 38.9 mL | Each |
| 15885364 | 330 mm | 15 mm | 38.9 to 53.0 mL | Each |
| 15815374 | 400 mm | 15 mm | 51.2 to 65.4 mL | Each |
| 15845374 | 500 mm | 15 mm | 68.9 to 83.1 mL | Each |
| 15875374 | 100 mm | 25 mm | 1.2 to 34.4 mL | Each |
| 15805384 | 150 mm | 25 mm | 19.6 to 58.9 mL | Each |
| 15835384 | 250 mm | 25 mm | 68.7 to 108.0 mL | Each |
| 15865384 | 330 mm | 25 mm | 108.0 to 147.3 mL | Each |
| 15895384 | 400 mm | 25 mm | 142.4 to 181.6 mL | Each |
| 15825394 | 500 mm | 25 mm | 191.4 to 230.7 mL | Each |
| 15855394 | 150 mm | 35 mm | 38.5 to 115.5 mL | Each |
| 15885394 | 250 mm | 35 mm | 134.7 to 211.7 mL | Each |
| 15815404 | 330 mm | 35 mm | 211.7 to 288.6 mL | Each |
| 15845404 | 400 mm | 35 mm | 279.0 to 356.0 mL | Each |
| 15875404 | 250 mm | 50 mm | 280.4 to 440.7 mL | Each |
| 15805414 | 330 mm | 50 mm | 440.7 to 600.9 mL | Each |
| 15835414 | 400 mm | 50 mm | 580.9 to 741.1 mL | Each |
| 15865414 | 500 mm | 50 mm | 781.2 to 941.4 mL | Each |
Product Code. 15825334
Brand: Diba 006EZ0625AF
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Includes
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting.
- PEEK™ column threads offer outstanding durability
- Precision borosilicate glass columns provide inertness, visibility, and performance
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Specifications
Specifications
| Temperature (Metric) Range | 4 to 40°C |
| Diameter (Metric) Inner | 6.6 mm |
| Type | Liquid Chromatography Column |
| Max. Pressure | 900 psi |
| Pore Size | 10 and 30 μm |
| Column Capacity | 4.8 to 7.5 mL |
| Length (Metric) | 250 mm |
| Material | Borosilicate Glass, PTFE, Polyethylene, FKM/FPM |
| Max. Temperature | 40°C |
| Includes | Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting. |