Learn More
Omeprazole-d3, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Omeprazole-d3 |
| CAS | 922731-01-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 2H3 H16 N3 O3 S |
| Quantity | 10 mg |
| Synonym | 1H-Benzimidazole','6-(methoxy-d3)-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]- (ACI), 6-(Methoxy-d3)-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole (ACI), 2-[(4-Methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-6-(trideuteriomethoxy)-1H-benzimidazole, 5-(Methoxy-d3)-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole','Omeprazole-d3, Omeprazole-D3 (benzimidazole-5-methoxy-D3) |
| InChI Formula | InChI=1S/C17H19N3O3S/c1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17/h5-8H,9H2,1-4H3,(H,19,20)/i3D3 |
| SMILES | [2H]C([2H])([2H])Oc1ccc2nc([nH]c2c1)S(=O)Cc3ncc(C)c(OC)c3C |
| IUPAC Name | 2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-6-(trideuteriomethoxy)-1H-benzimidazole |
| Molecular Weight (g/mol) | 348.43 |
| Show More |