Learn More
EVOS™ Olympus 4X Objective, super-apochromat, LWD
Click to view available options
Magnification Power:
4X
Unit Size:
Each
Description
- Has long working distance (LWD) objective that is optimized for imaging of slides, cell culture dishes and flasks, and microtiter plates
- Offers Apo correction from UV through IR ranges
- Parfocal for both visible and IR imaging
- Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
- Compatible with EVOS Cell Imaging systems
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Specifications
| Type | Objective Lens |
| Product Line | EVOS |
| Focus Distance | 13 mm |
| For Use With (Equipment) | EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Includes | Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates |
| Lens Type | Super-apochromat |
| Magnification Power | 4X |
| Numerical Aperture | 0.16 |
| Quantity | Each |
| Shipping Condition | Room Temperature |